missing translation for 'onlineSavingsMsg'
Learn More

Rhodium wire, 0.5mm (0.02in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 15415985
Click to view available options
Quantity:
7.5 cm
10 cm
30 cm
50 cm
250 cm
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 7.5 cm
Percent Purity 99.8%
Weight ≈2.44g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Rhodium wire, 0.5mm (0.02 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'