missing translation for 'onlineSavingsMsg'
Learn More

Rhodium powder, -325 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11318867
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
This item is not returnable. View return policy
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 0.5 g
UN Number UN3089
Percent Purity 99.95%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Rhodium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'