Alfa Aesar™ Rhodium(III) sulfate, bright plating solution, Rh 5% (cont. Rh)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 012635.NA

UNSPSC: 12141738

Code: NEW

Additional Details:
Additional Details: CAS Number: 10489-46-0 Weight: 0.01000kg


Quantity 2g

Product Code. 11410310

Quantity Price
1 £ 633.46 / 2g
Estimated Shipment date
from Supplier 18-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Rhodium(III) sulfate
CAS 10489-46-0
Name Note Bright plating solution
Linear Formula Rh2(SO4)3
Formula Weight 493.99
MDL Number MFCD00016309
UN Number UN3264

Specification

Formula Weight 493.99
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number FF9335000
Health Hazard 1 Danger
Linear Formula Rh2(SO4)3
Density 1.4g/mL
CAS 10489-46-0
Chemical Name or Material Rhodium(III) sulfate
MDL Number MFCD00016309
Physical Form Liquid
UN Number UN3264
Odor Acidic
Color Red-orange
Name Note Bright plating solution
Concentration Rh 5% (cont. Rh)
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA TSCA
Quantity 2g
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.