missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) sulfate, bright plating solution, Rh 5% (cont. Rh), Thermo Scientific™

Catalog Number p-7049065
Click to view available options
Quantity:
0.5 g
This item is not returnable. View return policy
10489-46-0
O12Rh2S3
493.98
MFCD00016309
YWFDDXXMOPZFFM-UHFFFAOYSA-H
rhodium iii sulfate solution, rhodium 3+ trisulfate, dirhodium trisulphate, rhodium sulfate, rhodium sulfate solution, acmc-1bntj, sulfuric acid, rhodium 3+ salt 3:2, rhodium iii sulfate, solution, dirhodium 3+ trisulfate, sulfuric acid,rhodium 3+ salt 3:2
159290
rhodium(3+);trisulfate
[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10489-46-0
O12Rh2S3
493.98
MFCD00016309
YWFDDXXMOPZFFM-UHFFFAOYSA-H
rhodium iii sulfate solution, rhodium 3+ trisulfate, dirhodium trisulphate, rhodium sulfate, rhodium sulfate solution, acmc-1bntj, sulfuric acid, rhodium 3+ salt 3:2, rhodium iii sulfate, solution, dirhodium 3+ trisulfate, sulfuric acid,rhodium 3+ salt 3:2
159290
rhodium(3+);trisulfate
[Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Red to Orange
Linear Formula Rh2(SO4)3
UN Number UN3264
Concentration Rh 5% (cont. Rh)
Density 1.4 g/mL
Solubility Information Miscible with water,ethanol and with a 3:1 ethanol/water mixture.
Formula Weight 493.99
Odor Acidic
Quantity 0.5 g
Physical Form Liquid
Chemical Name or Material Rhodium(III) sulfate, Bright plating solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'