Learn More
Rhodium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Rhodium(III) chloride hydrate is used as a catalyst for the conversion of methane to acetic acid, isomerization of alkenes, reduction of aromatic rings and hydration of acetylene. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals. It plays an important role as a catalyst for conjugate reduction of cinnamaldehydes followed by cross-coupling with arylboronic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100°C (decomposition) |
Color | Red |
Physical Form | Crystalline |
Quantity | 1 g |
Merck Index | 14,8188 |
Solubility Information | Soluble in water and alcohol. Slightly soluble in acetone. |
Formula Weight | 209.26 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Premion™ |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only