Learn More
Alfa Aesar™ Rhodium(II) octanoate dimer
Brand:  Alfa Aesar™ 039825.06
Code : NEW
Additional Details : CAS Number : 73482-96-9 Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 11329147
  • £945.90 / 5g
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Chemical Identifiers
Rhodium(II) octanoate dimer
[Rh[CH3(CH2)6CO2]2]2
MFCD00064724
73482-96-9
778.63
Specifications
Rhodium(II) octanoate dimer
Powder
778.63
Soluble in hot alcohol, dichloromethane, toluene and acetic acid, slightly soluble in alcohol
5g
73482-96-9
[Rh[CH3(CH2)6CO2]2]2
MFCD00064724
Green