missing translation for 'onlineSavingsMsg'
Learn More

Rhodium, 5% on carbon paste, Type 594, Thermo Scientific™

Catalog Number 15447767
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]
This item is not returnable. View return policy
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
23948
CHEBI:33359
rhodium
[Rh]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Gray to Black
Physical Form Solid
Quantity 25 g
Odor Odorless
Chemical Name or Material Rhodium, 5% on carbon paste, Type 594
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'