missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Rhodium, 5% on activated carbon powder, Type C101038-5, reduced, nominally 50% water wet

Brand:  Alfa Aesar™ 011761.18

Code : NEW

Additional Details : CAS Number : 7440-16-6 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11361828

  • £1180.80 / 50g


Estimated Shipment date
from Supplier 23-04-2018
Add to basket

Specifications

Chemical Identifiers

Rhodium
7440-16-6
5% on activated carbon powder, Type C101038-5, Reduced, nominally 50% water wet
MFCD00011201

Specifications

Rhodium
7440-16-6
MFCD00011201
50g
5% on activated carbon powder, Type C101038-5, Reduced, nominally 50% water wet
Powder
Black
Odorless
Videos
SDS