Learn More
Thermo Scientific Alfa Aesar Rhodium 10%, Palladium 1% on activated carbon powder
Brand: Thermo Scientific Alfa Aesar 044124.22
Rhodium 10%, Palladium 1% on activated carbon powder
Code : NEW
Additional Details : Weight : 0.10000kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III