Learn More
Thermo Scientific Alfa Aesar Rhodium, 0.5% on 3mm (0.12in) alumina pellets
Brand: Thermo Scientific Alfa Aesar 042507.09
Rhodium, 0.5% on 3mm (0.12in) alumina pellets
Code : NEW
Additional Details : CAS Number : 7440-16-6 Weight : 0.01000kg