Learn More
Thermo Scientific Alfa Aesar Rhodium, 0.5% on 1mm (0.040in) alumina spheres
Brand: Thermo Scientific Alfa Aesar 045639.09
Rhodium, 0.5% on 1mm (0.040in) alumina spheres
Code : NEW
Additional Details : CAS Number : 7440-16-6 Weight : 0.01000kg