missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fc Receptor Binding Inhibitor anti-Rhesus Monkey, Polyclonal, eBioscience™

Polyclonal Antibody

Brand:  Invitrogen™ 14-9165-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15577586

  • £140.00 / 100 tests

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!

Alert:

Contains Sodium azide


Specifications

Specifications

5μL (10μg)/test
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2° to 8°C
Flow Cytometric Analysis
100 tests
Rhesus
Videos
Documents

For Research Use Only