Learn More
Thermo Scientific™ Rheodyne™ Sample Loops
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Brand:  Thermo Scientific™ 7022
Code : 16
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10591305
  • £46.40 / Each
Estimated Shipment
18-05-2018
Add to basket
Specifications
Specifications
Rheodyne 7010/7125
For Rheodyne 7010/7125; 20uL; 0.30mm ID
20μL
Sample Loop
0.30mm
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.