missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(VII) oxide, 99.99% (metals basis) , Alfa Aesar™

Moisture Sensitive; Sublimation temperature/start: 250°C (482°F)

Brand:  Alfa Aesar 011339.09

Code : NEW

Additional Details : CAS Number : 1314-68-7 Weight : 0.01000kg Transport : UN number : 3085 Chem class : 1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11391099

  • POA / 10g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

Rhenium(VII) oxide
Crystalline
O7Re2
MFCD00011199
1314-68-7
99.99% (metals basis)
484.4

Specifications

Rhenium(VII) oxide
Crystalline
O7Re2
484.4
Odorless
10g
1314-68-7
99.99% (metals basis)
Yellow powder
MFCD00011199
White
Videos
SDS