missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium(VI) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
455.35 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Rhenium(VI) oxide is involved as a catalyst for the hydrogenation of carboxylic acids and carboxamides. It is involved in the alkene epoxidation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 400°C (decomposition) |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 236.2 |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Rhenium(VI) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'