missing translation for 'onlineSavingsMsg'
Learn More

Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis), Thermo Scientific Chemicals

Catalog Number 11311408
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-15-5
Re
186.21
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
23947
CHEBI:49882
rhenium
[Re]
This item is not returnable. View return policy
7440-15-5
Re
186.21
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
23947
CHEBI:49882
rhenium
[Re]

Form: Approx. 3.29g/25 x 25mm; Solid in various forms

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver gray foil
Assay Percent Range 99.97% (Metals basis)
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Chemical Name or Material Rhenium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'