missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGM-A Goat anti-Chicken, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | RGM-A |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant chicken RGM-A Trp30-Asn404 (Asn404Asp) Accession # Q8JG54 |
Gene ID (Entrez) | 56963 |
Gene Accession No. | Q8JG54 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
RGM-A Polyclonal specifically detects RGM-A in Chicken samples. It is validated for Western Blot.Specifications
RGM-A | |
56963 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects chicken RGM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human RGM and recombinant mouse RGM. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant chicken RGM-A Trp30-Asn404 (Asn404Asp) Accession # Q8JG54 | |
Q8JG54 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only