Revco™ High-Performance Lab Freezers

Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers.

Overview
Brand: Thermo Scientific Revco

Manufacturer Part Number: ULT1230W

UNSPSC: 41103011

Code: 77

Additional Details:
Additional Details: Weight: 127.00000kg



Product Code. 12938884

Quantity Price
1 £ 3305.0 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Certifications/Compliance UL and CUL listed. CE marked.
Amperage 8A
Capacity 11.5 cu. ft., 326L
Breaker 15A
Cabinet Type Space Saver
Capacity (Metric) 326L
Defrost Automatic
Depth (Metric) Interior 53.3cm
Dimensions (L x W x H) Exterior 29.3 x 24 x 73.6 in. (74.3 x 61 x 186.9cm)
Dimensions (D x W x H) Interior 21 x 20 x 52.4 in. (53.3 x 50.8 x 133.1cm)
Height (Metric) Exterior 186.9cm
Height (Metric) Interior 133.1cm
Hertz 50Hz
Length (Metric) Exterior 74.3cm
Plug Type U.K.
Shelves 4
Refrigerant CFC-free
Shipping Weight (Metric) 127kg
Temperature Control Microprocessor
Temperature Range (Metric) -30°C
Type High-Performance Freezer
Voltage 230V
Width (Metric) Exterior 61cm
Doors Single, Solid
Width (Metric) Interior 50.8cm
Electrical Requirements 230V 50Hz
Warranty Warranty varies by country, please contact us for details

  • Temperature factory-preset at -20° or -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-30°C Freezers

  • Automatic defrost and condensate removal
  • ensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

-20°C Freezers

  • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
  • Gravity convection air system for maximum stability without a blower or fan
  • Manual defrost

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Documentation