missing translation for 'onlineSavingsMsg'
Learn More

Revco™ High-Performance Lab Freezers

Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ High-Performance Lab Freezers.

Brand:  Thermo Scientific Revco ULT430W

Code : 77

Additional Details : Weight : 96.00000kg

 View more versions of this product

Product Code. 12928884

  • £3293.07 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Temperature factory-preset at -20° or -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-30°C Freezers

  • Automatic defrost and condensate removal
  • ensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

-20°C Freezers

  • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
  • Gravity convection air system for maximum stability without a blower or fan
  • Manual defrost

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Specifications

Specifications

6A
Undercounter
86.4cm
50Hz
CFC-free
96kg
61cm
Automatic
139L
20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8cm)
−30°C
230V
230V 50Hz
15A
50.8cm
52.1cm
73.4cm
3
Microprocessor
50.8cm
UL and CUL listed. CE marked.
28.9 x 24 x 34 in. (73.4 x 61 x 86.4cm)
UK
High-Performance Freezer
Single, Solid
Warranty varies by country, please contact us for details
Videos
Documentation Instrument & Equipment Services