missing translation for 'onlineSavingsMsg'
Learn More

RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors

For fine grinding of soft to medium-hard and fibrous materials.

Brand:  RETSCH 02.608.0042

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10742171

  • £1560.00 / Each
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Accessory for use with ZM 200 Ultra Centrifugal Mill
  • Wear resistant coating
Specifications

Specifications

Stainless Steel
6,000 to 18,000 rpm
Centrifugal Mill
24 Tooth SS Push Fit
9.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers