Learn More
RETSCH Test Sieves - 13 Mesh or Less
Stainless steel with wire gauze
£110.00 - £157.00
Specifications
Diameter (Metric) Outer | 8 in. |
---|---|
Mesh Material | Stainless Steel |
For Use With (Application) | Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. |
Depth (English) | 2 in. |
Depth (Metric) | 5.08cm |
Product Code | Brand | Pore Size | Tyler Standard | U.S. Standard | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Tyler Standard | U.S. Standard | Price | Quantity & Availability | |||
10390762
|
RETSCH
60.150.001700 |
1.7mm | 12 Mesh | No. 12 |
£110.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10075011
|
RETSCH
60.150.002000 |
2mm | 10 Mesh | No. 10 |
£134.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
10710283
|
RETSCH
60.150.002360 |
2.36mm | 8 Mesh | No. 8 |
£110.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10218931
|
RETSCH
60.150.002800 |
2.8mm | 7 Mesh | No. 7 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10497001
|
RETSCH
60.150.005600 |
5.6mm | 3.5 Mesh | No. 3.5 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10497191
|
RETSCH
60.150.006300 |
6.3mm | 0.25 Mesh | No. 1.4 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10136171
|
RETSCH
60.150.003350 |
3.35mm | 6 Mesh | No. 6 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10146171
|
RETSCH
60.150.004750 |
4.75mm | 4 Mesh | No. 4 |
£135.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
10407201
|
RETSCH
60.150.006700 |
6.7mm | 0.26 Mesh | No. 0.26 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10350862
|
RETSCH
60.150.004000 |
4mm | 5 Mesh | No. 5 |
£135.00
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
10701243
|
RETSCH
60.150.008000 |
8mm | 0.3 Mesh | No. 2.5 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10477771
|
RETSCH
60.150.012500 |
12.5mm | 0.5 Mesh | No. 0.5 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10477961
|
RETSCH
60.150.013200 |
13.2mm | 0.53 Mesh | No. 0.53 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10418161
|
RETSCH
60.150.016000 |
16mm | 0.62 Mesh | No. 0.62 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10525072
|
RETSCH
60.150.009500 |
9.5mm | 0.37 Mesh | No. 0.37 |
£135.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10751813
|
RETSCH
60.150.011200 |
11.2mm | 0.43 Mesh | No. 0.43 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10418351
|
RETSCH
60.150.019000 |
19mm | 0.75 Mesh | No. 0.75 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10428351
|
RETSCH
60.150.022400 |
22.4mm | 0.87 Mesh | No. 0.87 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10600462
|
RETSCH
60.150.045000 |
45mm | 1.75 Mesh | No. 1.75 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10095011
|
RETSCH
60.150.037500 |
37.5mm | 1.5 Mesh | No. 1.5 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10278591
|
RETSCH
60.150.026500 |
26.5mm | 1.06 Mesh | No. 1.06 |
£151.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10095001
|
RETSCH
60.150.031500 |
31.5mm | 1.25 Mesh | No. 1.25 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10238691
|
RETSCH
60.150.053000 |
53mm | 2.12 Mesh | No. 2.12 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10426241
|
RETSCH
60.150.050000 |
50mm | 2 Mesh | No. 2 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10146881
|
RETSCH
60.150.063000 |
63mm | 2.5 Mesh | No. 2.5 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10340672
|
RETSCH
60.150.075000 |
75mm | 3 Mesh | No. 3 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10650292
|
RETSCH
60.150.090000 |
90mm | 3.5 Mesh | No. 3.5 |
£157.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
Description
Dimensions: 8 dia. × 2 in.D (20.3 × 5cm)
Compliance
Meet ASTM E11
Specifications
8 in. | |
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. | |
5.08cm | |
ISO 3310 | |
Stainless Steel |
Stainless Steel | |
2 in. | |
Test Sieve | |
8 in. |