Learn More
RETSCH Economy Clamping Device for AS 200 Sieve Shaker
Provides reliable, quick and convenient clamping of sieves in dry operation.
Brand: RETSCH 32.662.0003
Additional Details : Weight : 3.60000kg
Description
- Threaded rods and clamping nuts
- Stainless-steel cover
Specifications
Test Sieves,100 to 203 mm dia. | |
Economy Clamping Device | |
Reliable, quick, convenient clamping of sieves in dry operation | |
AS 200 sieve shaker |