missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Straight Inlet Liners for Splitless Injection
Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition
Brand: Restek™ 22402
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
- Compatible with Agilent™/Finnigan™ columns
- Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples; also, packing in wool is recommended to aid in vaporization of high molecular weight compounds and to minimize discrimination
- O.D. × Length: 6.5 × 78.5mm
Specifications
Glass Splitless with Wool | |
IP Splitless Straight Inlet Liner | |
7.85 cm | |
0.65 cm | |
Borosilicate Glass |
Deactivated Wool | |
25/Pk. | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |