missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Straight Inlet Liners for Splitless Injection

Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition

Brand:  Restek™ 20773

Code : Z3

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 12922749

  • £96.80 / Pack of 5
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

  • Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
  • Compatible with Agilent™/Finnigan™ columns
  • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples; also, packing in wool is recommended to aid in vaporization of high molecular weight compounds and to minimize discrimination
  • O.D. × Length: 6.5 × 78.5mm

Specifications

Specifications

Glass Splitless
5/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
IP Splitless Straight Inlet Liner
7.85 cm
0.65 cm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers