Learn More
Restek™ Rtx™-1 Capillary Columns
Similar in polarity to DB-1 and HP-1
£680.02 - £813.12
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Temperature Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Temperature Range | Price | Quantity & Availability | ||
12925309
|
Restek™
10108 |
0.25 mm | 0.10 μm | 30 m | -60°C to +330/350°C |
£696.96
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||
12975309
|
Restek™
10109 |
0.32 mm | 0.1 μm | 30 m | -60°C to +330/350°C |
£739.31
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12995339
|
Restek™
10123 |
0.25 mm | 0.25 μm | 30 m | -60°C to +330/350°C |
£680.02
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12965349
|
Restek™
10124 |
0.32 mm | 0.25 μm | 30 m | -60°C to +330/350°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12935409
|
Restek™
10139 |
0.32 mm | 0.5 μm | 30 m | -60°C to +330/350°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12965449
|
Restek™
10153 |
0.25 mm | 1 μm | 30 m | -60°C to +320/340°C |
£680.02
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12915459
|
Restek™
10154 |
0.32 mm | 1 μm | 30 m | -60°C to +320/340°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
10430847
|
Restek™
10154 |
0.32 mm | 1 μm | 30 m | -60°C to +320/340°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12948119
|
Restek™
10170 |
0.53 mm | 1.5 μm | 30 m | -60°C to +310/330°C |
£813.12
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12938149
|
Restek™
10178 |
0.32 mm | 5 μm | 30 m | -60°C to +260/280°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12988149
|
Restek™
10179 |
0.53 mm | 5 μm | 30 m | -60°C to +270/290°C |
£813.12
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
10116533
|
Restek™
10184 |
0.32 mm | 3 μm | 30 m | -60°C to +280/300°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12918189
|
Restek™
10184 |
0.32 mm | 3 μm | 30 m | -60°C to +280/300°C |
£724.79
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12978189
|
Restek™
10185 |
0.53 mm | 3 μm | 30 m | -60°C to +270/290°C |
£813.12
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
10087611
|
Restek™
10155 |
0.53 mm | 1 μm | 30 m | -60°C to +310/330°C |
£813.12
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
12918079
|
Restek™
10155 |
0.53 mm | 1 μm | 30 m | -60°C to +310/330°C |
£813.12
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||
Description
- Nonpolar; coated with crossbonded polydimethylsiloxane (100% dimethylpolysiloxane)
- Manufactured from high-quality fused silica tubing
- Surrounded by a rugged stainless-steel cage
- Excellent thermal stability for low bleed at high temperatures
Solvents, Petroleum products, Pharmaceuticals, Waxes, Fuel oils
Specifications
Rtx™-1 | |
G1, G2, G38 | |
Fused Silica |