Learn More
Restek™ Normal Phase Resprep SPE Cartridges
Brand: Restek™ 24032
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Manufactured using high density polypropylene
- May be processed by any of these techniques: positive pressure, sidearm flask, centrifuge or vacuum manifold
Specifications
Hydrophilic (Polar) | |
Normal | |
SPE Cartridge | |
HDPE, Polyethylene Frits |
Florisil PTFE Frits | |
Used to Extract Hydrophilic Analytes From Non-Polar Matrices, Such as Organic Solvents (e.g., Polar Contaminants From Sample Extracts) | |
500 mg/3 mL | |
3 mL, 500 mg |