Learn More
Restek™ Injector/Detector Plug Nut
Cap off or check injector or detector for leaks, cleaning, flow-rate.
Brand: Restek™ 21883
Additional Details : Weight : 1.00000kg
Description
- Use to cap off an injector to isolate leaks
- Use to cap off a detector for thermal cleaning
- Use to check a detector or make-up gas flow rate
- Use to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base
Specifications
Nut | |
Cap Off or Check Injector or Detector for Leaks, Cleaning, Flow-Rate. | |
2/Pk. | |
HP GC |