missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Convenience Kits with Vials: Untreated
Glass vials come standard in these easy-to-use convenience kits
Brand: Restek™ 21195
This item is not returnable.
View return policy
Description
- Vials packaged in a clear-lid tray
- Preassembled caps with septa packaged separately in a plastic bag
Specifications
Amber | |
Polypropylene | |
1000 Pk. |
2 mL | |
Screw Cap, Open Top | |
8 mm |