missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ 20 mm Aluminum Seals with Septa, Assembled
Preassembled seals save time and increase productivity
£100.80 - £1017.60
Product Code | Brand | Septum | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Septum | No. per Pack | Price | Quantity & Availability | ||||
13129662
|
Restek™
21761 |
PTFE, Butyl Rubber | 100 |
£114.00
Pack of 100 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
13139662
|
Restek™
21762 |
PTFE, Butyl Rubber | 1000 |
£1017.60
Pack of 1000 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
10698904
|
Restek™
21763 |
PTFE, Silicone | 100 |
£100.80
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
13149662
|
Restek™
21763 |
PTFE, Silicone | 100 |
£100.80
Pack of 100 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
13159662
|
Restek™
21764 |
PTFE, Silicone | 1000 |
£870.00
Pack of 1000 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
Description
- Color: Silver
- Material: PTFE/Gray Butyl Rubber Septa
Specifications
Silver | |
Aluminum | |
Crimp Cap | |
20 mm |