missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Resistop Gloves

Comfortable and sturdy polyamide-cotton knit construction gives these gloves thermal and mechanical resistance. Honeywell Safety Products™ Resistop Gloves are non-pilling and suitable for precision work.

Brand:  Honeywell Safety Products™ RGT099T8

Code : NEW

Additional Details : Weight : 0.08100kg

 View more versions of this product

Product Code. 12663383

  • £6.80 / Each


Estimated Shipment
04-05-2021
Add to basket

Description

Description

  • Suitable for handling sharp and hot objects.
  • Non-pilling knit polyamide exterior.
    • Abrasion, cutting, thermal resistance.
    • Suitable for precision work.
    • Conforms to hand.
  • Cotton interior for comfort and sweat absorption.
  • Thick construction absorbs shocks.
  • White color enhances hygiene and cleanliness.
Specifications

Specifications

Dexterity: 5
Handling of sharp parts in automotive and mechanical industries. Assembling operations, removal of molds up to 100°C for 30 seconds maximum. Automotive, Aviation, Glass, Logistics and Offshore industries.
Polyamide, Cotton
8
10Pair/PK
25.5 cm
Knitted Polyamide/Cotton
6cm
Cuts,Heat
4
White
CATEGORY CE II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number N°0075/797/162/09/08/0345, EC Attestation Number N°0075/797/162/09/08/0345
Videos
SDS
Documents