missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Resistop Gloves
Click to view available options
Size:
8
9
Description
- Suitable for handling sharp and hot objects.
- Non-pilling knit polyamide exterior.
- Abrasion, cutting, thermal resistance.
- Suitable for precision work.
- Conforms to hand.
- Cotton interior for comfort and sweat absorption.
- Thick construction absorbs shocks.
- White color enhances hygiene and cleanliness.
Specifications
Certifications/Compliance | CATEGORY CE II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number N°0075/797/162/09/08/0345, EC Attestation Number N°0075/797/162/09/08/0345 |
Classification | Dexterity: 5 |
Description | Knitted Polyamide/Cotton |
Color | White |
Cut Resistance Rating | 3 |
Material | Polyamide, Cotton |
Product Type | Gloves |
Size | 8 |
Temperature (Metric) Max | 100°C |
For Use With (Application) | Handling of sharp parts in automotive and mechanical industries. Assembling operations, removal of molds up to 100°C for 30 seconds maximum. Automotive, Aviation, Glass, Logistics and Offshore industries. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'