missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research™ plus EU-IVD 8-Channel Variable Volume Pipette

The ultra light Eppendorf Research plus pipette meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.

Brand:  Eppendorf™ 3122000051

Code : 97

Additional Details : Weight : 0.63000kg

 View more versions of this product

Product Code. 10172494

  • £737.24 / Each


Estimated Shipment: 25-02-2022
Add to basket

Description

Description

The ultra light Eppendorf Research plus pipette meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.

  • Air-displacement pipette for accurate pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
  • Spring loaded tip cone (not available for 5 mL and 10 mL pipettes) for minimal tip attachment forces helps to reduce stress
  • Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
  • Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs to ensure contamination-free operation
Specifications

Specifications

EU-IVD
epT.I.P.S.™ 300 μL
Orange
8
30 to 300 μL
Forward pipetting, Reverse pipetting, Removal of supernatants, Sample mixing, Phase extraction, Volume determination, Filling of plates, gels, and reaction vessels
Autoclavable
No
Multichannel manual pipette
Videos
SDS
Documents
Certificates
View accessory products