missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ SilTite™ FingerTite Ferrule
For gas chromatography systems
£70.80
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
15238983
|
Trajan™
073632 |
0.7 mm |
£70.80
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15228983
|
Trajan™
073631 |
0.5 mm |
£70.80
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15218983
|
Trajan™
073630 |
0.4 mm |
£70.80
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
GC Ferrule | |
10/Pk. |