missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for All-glass Filter Holder Kits, 90 mm
Accessories for filter holders for sample preparation.
£169.00 - £585.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Sterility | Non-sterile |
Diameter (Metric) Inside | 90 mm |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10769201
|
Merck
XX1009002 |
Screen |
£291.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15844166
|
Merck
XX1019004 |
Vacuum Base and Cap |
£585.00
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10791073
|
Merck
XX1009010 |
Gasket |
£169.00
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
15824166
|
Merck
XX1019000 |
Funnel |
£303.00
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
15834166
|
Merck
XX1019003 |
Spring Clamp, Aluminum |
£382.00
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
Specifications
Not Autoclavable | |
Non-sterile | |
90 mm |