missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Replacement Needle and Plunger Kit for PerkinElmer Autosampler Syringes
Repair kit for PerkinElmer Autosystem™ and Clarus™ Removable Needle Autosampler Syringes
Brand: Trajan Scientific and Medical™ 033750
Code : 13
Additional Details : Weight : 0.50000kg
Description
Includes:
- 70mm cone tipped needle
- 0.5μL volume
Choose Needle:
- 26 gauge, 0.47 OD
- 23 gauge, 0.63 OD
Specifications
26 G | |
Replacement Needle and Plunger Kit | |
Cone |
0.47 mm | |
0.5 μL | |
AutoSystem and Clarus Removable Needle Syringe |