Learn More
SGE™ Repeating Adapter
Improve precision and reproducibility when repeatedly dispensing the same volume manually. SGE Analytical Science™ Repeating Adaptors are suitable for syringes with capacities from 500nL through to 500μL.
Brand: SGE™ 031930
Code : 13
Additional Details : Weight : 0.03384kg
Description
- Can be preset to any desired volume
- Helps prevent plunger blow out at elevated pressures
- Repeating adapter can be fitted to manually operated syringes
Specifications
Helps prevent plunger blow out at elevated pressures | |
Manually Operated Syringe |