missing translation for 'onlineSavingsMsg'
Learn More

Remoxipride hydrochloride, Tocris Bioscience™

Selective D2-like antagonist

Brand:  Tocris (Bio-Techne) 0916/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13468153

  • £401.31 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dopamine D2 receptor antagonist showing selectivity over D3 and D4 receptors (Ki values are ∽ 300, ∽ 1600, and ∽ 2800 nM for D2, D3 and D4 receptors respectively). Exhibits antipsychotic activity in vivo with no extrapyramidal side effects. 50-fold more potent than sulpiride in antagonising the effects of apomorphine in the rat.
Specifications

Chemical Identifiers

73220-03-8
407.733
remoxipride hydrochloride, roxiam, remoxipride hcl, a 33547 hydrochloride, unii-qs4s72u30k, s-remoxipride, remoxipride hydrochloride anhydrous, s---2-3-bromo-2,6-dimethoxybenzamido methyl-1-ethylpyrrolidine hydrochloride, s---3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl benzamide hydrochloride, benzamide, 3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl-, hydrochloride, s--
3-bromo-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dimethoxybenzamide;hydrochloride
C16H24BrClN2O3
WCPXLMIPGMFZMY-MERQFXBCSA-N
15565709
CCN1CCCC1CNC(=O)C2=C(C=CC(=C2OC)Br)OC.Cl

Specifications

50mg
73220-03-8
remoxipride hydrochloride, roxiam, remoxipride hcl, a 33547 hydrochloride, unii-qs4s72u30k, s-remoxipride, remoxipride hydrochloride anhydrous, s---2-3-bromo-2,6-dimethoxybenzamido methyl-1-ethylpyrrolidine hydrochloride, s---3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl benzamide hydrochloride, benzamide, 3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl-, hydrochloride, s--
CCN1CCCC1CNC(=O)C2=C(C=CC(=C2OC)Br)OC.Cl
407.733
407.73
Remoxipride hydrochloride
C16H24BrClN2O3
WCPXLMIPGMFZMY-MERQFXBCSA-N
3-bromo-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dimethoxybenzamide;hydrochloride
15565709
>98%
Videos
Documents