missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Removable Needle for Agilent Autosampler Syringes
Quality needle replacements for use with Agilent autosampler syringes with removable needle.
Brand: Trajan Scientific and Medical™ 037717
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Needle length is the exposed length of needle
- For use with Agilent Technologies autosamplers
Specifications
23 G | |
Replacement Needle | |
Cone |
0.63 mm | |
50 mm | |
Syringes (10μL) |