Learn More
Thermo Scientific™ Remel™ BactiDrop Lactophenol Aniline Blue
Remel BactiDrop Lactophenol Aniline Blue is for staining fungal elements.
Brand: Thermo Scientific™ R21526
Code : 50
Additional Details : CAS Number : 56-81-5 Weight : 0.05000kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II
Description
Remel BactiDrop Lactophenol Aniline Blue is for staining fungal elements.
Ordering Alerts: Supplied in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
Specifications
0.75 mL/Ampule | |
Lactophenol Aniline Blue | |
50 Pk. |
Safety and Handling
- Danger
Hazard Category
- Germ cell mutagenicity Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H341-Suspected of causing genetic defects.
- H331-Toxic if inhaled.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- H373-May cause damage to organs.
- H314-Causes severe skin burns and eye damage.