Learn More
Remacemide hydrochloride, Tocris Bioscience™
NMDA antagonist; blocks ion channel and allosteric modulatory site
Brand:  Tocris (Bio-Techne) 162250
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13439823
  • £374.85 / 50mg
Estimated Shipment date
from Supplier 02-03-2020
Description
Description
Non-competitive NMDA receptor antagonist; blocks ion channel and allosteric modulatory site (IC50 = 8 - 68 mM). Anticonvulsant in vivo and metabolizes to a more potent desglycine analog. Weakly blocks voltage-dependent Na+ channels (IC50 = 161 mM).
Specifications
Chemical Identifiers
111686-79-4
304.818
remacemide hydrochloride, remacemide hcl, remacemide hydrochloride usan, 2-amino-n-1,2-diphenylpropan-2-yl acetamide hydrochloride, +--2-amino-n-1-methyl-1,2-diphenylethyl acetamide monohydrochloride, 2-amino-n-1-methyl-1,2-diphenylethyl acetamide hydrochloride, acetamide, 2-amino-n-1-methyl-1,2-diphenylethyl-, +/--cas, dsstox_cid_25683, dsstox_rid_81056, dsstox_gsid_45683
2-amino-N-(1,2-diphenylpropan-2-yl)acetamide;hydrochloride
C17H21ClN2O
HYQMIUSWZXGTCC-UHFFFAOYSA-N
60510
CC(CC1=CC=CC=C1)(C2=CC=CC=C2)NC(=O)CN.Cl
Specifications
50mg
111686-79-4
remacemide hydrochloride, remacemide hcl, remacemide hydrochloride usan, 2-amino-n-1,2-diphenylpropan-2-yl acetamide hydrochloride, +--2-amino-n-1-methyl-1,2-diphenylethyl acetamide monohydrochloride, 2-amino-n-1-methyl-1,2-diphenylethyl acetamide hydrochloride, acetamide, 2-amino-n-1-methyl-1,2-diphenylethyl-, +/--cas, dsstox_cid_25683, dsstox_rid_81056, dsstox_gsid_45683
CC(CC1=CC=CC=C1)(C2=CC=CC=C2)NC(=O)CN.Cl
304.818
304.82
Remacemide hydrochloride
C17H21ClN2O
HYQMIUSWZXGTCC-UHFFFAOYSA-N
2-amino-N-(1,2-diphenylpropan-2-yl)acetamide;hydrochloride
60510
>99%