missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Relaxin-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Relaxin-1 |
---|---|
Clone | 361505 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Relaxin-1 Monoclonal specifically detects Relaxin-1 in Human samples. It is validated for Western Blot.Specifications
Relaxin-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
P04808 | |
6013 | |
E. coli-derived recombinant human Relaxin-1 Lys26-Cys185 Accession # P04808 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects both the mature form and the proform of human Relaxin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Relaxin-2 or rhRelaxin-3 is observed. |
361505 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, Relaxin1, RLN1, RLXH1 | |
RLN1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only