Learn More
Reinnervate Alvetex™ Scaffold Multiwell Plate Formats
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
Brand:  Alvetex AVP002-320
UNSPSC : 41122102
Code : NEW
Additional Details : Weight : 27.20000kg
 View more versions of this product
Product Code. 15216700
  • POA / Pack of 320
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
12
Alvetex™ (porous polystyrene scaffold)
320
320 x 12 well plates