Reinnervate Alvetex™ Scaffold Multiwell Plate Formats
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
Overview
Brand: Alvetex
Manufacturer Part Number: AVP010-10
ALVETEX 384-WELL PLATE, 10 PLATES
UNSPSC: 41122102
Code: NEW
Additional Details:
Additional Details: Weight: 0.95000kg
Product Code. 15284196
Quantity Price
1 £ 845.91 / Pack of 10
Estimated Shipment
04-04-2017
Add to basket
Description and Specification
Specification
Material Alvetex™ (porous polystyrene scaffold)
No. per Pack 10 x 384 well plates
Quantity 10
No. of Wells 384
Documentation
Videos