Reinnervate Alvetex™ Scaffold Multiwell Plate Formats
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
Overview
Brand: Alvetex
Manufacturer Part Number: AVP009-80
ALVETEX 96-WELL PLATE, 80 PLATES FOR 3D
UNSPSC: 41122102
Code: NEW
Additional Details:
Additional Details: Weight: 6.80000kg
Product Code. 15266700
Quantity Price
1 £ 5719.4400000000005 / Pack of 80
Estimated Shipment
03-03-2017
Add to basket
Description and Specification
Specification
No. of Wells 96
Quantity 80
No. per Pack 80 x 96 well plates
Material Alvetex™ (porous polystyrene scaffold)
Documentation
Videos