Learn More
Reinnervate Alvetex™ Scaffold Multiwell Plate Formats
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
Brand:  Alvetex AVP009-320
UNSPSC : 41122102
Code : NEW
Additional Details : Weight : 27.20000kg
 View more versions of this product
Product Code. 15276700
  • POA / Pack of 320
Estimated Shipment
04-08-2017
Add to basket
Specifications Description & Specifications
Specifications
96
Alvetex™ (porous polystyrene scaffold)
320
320 x 96 well plates