Reinnervate Alvetex™ Scaffold Multiwell Plate Formats
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
Overview
Brand: Alvetex
Manufacturer Part Number: AVP006-320
UNSPSC: 41122102
Code: NEW
Additional Details:
Additional Details: Weight: 27.20000kg
Product Code. 15246700
Quantity Price
1 £ 9562.18 / Pack of 320
Estimated Shipment
13-06-2017
Add to basket
Description and Specification
Specification
Material Alvetex™ (porous polystyrene scaffold)
Quantity 320
No. per Pack 320 x 24 well plates
No. of Wells 24
Documentation
Videos