Learn More
Reichert™ Reichert™ Goldberg Ultra-Precision Handheld Refractometers
Offer reading accuracy better than most benchtop laboratory refractometers
Brand:  Reichert™ 13104190
Code : 04
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11943365
  • £1444.53 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Distilled Water
0 to 160ppt
25.4mm
0.0002nD/2ppt
1.33 to 1.37
25.4mm
±0.3ppt
Ideal for use in quality control, research and clinical laboratories
Soft holster case
0.0001
157.4mm
6.2 x 1 x 1 in. (157.4 x 25.4 x 25.4mm)
Videos
Documentation View accessory products