missing translation for 'onlineSavingsMsg'
Learn More

Reichert™ Goldberg Ultra-Precision Handheld Refractometers

Offer reading accuracy better than most benchtop laboratory refractometers

Brand:  Reichert Instruments 13104230

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11930917

  • £1444.53 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Distilled Water
0 to 30%
25.4mm
0.1%
1.33 to 1.37
25.4mm
±0.1%
Ideal for use in quality control, research and clinical laboratories
Soft holster case
0.1
157.4mm
6.2 x 1 x 1 in. (157.4 x 25.4 x 25.4mm)
Videos
Documentation View accessory products