missing translation for 'onlineSavingsMsg'
Learn More

Mouse Reg1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications

POA

Specifications

Antigen Reg1
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Description

Description

Reg1 Polyclonal specifically detects Reg1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Reg1
Western Blot 0.1 ug/mL
Polyclonal
Sheep
Mouse
ICRF, Islet cells regeneration factor, Islet of Langerhans regenerating protein, lithostathine-1-alpha, Pancreatic stone protein, pancreatic stone protein, secretory, Pancreatic thread protein, protein-X, PSPregenerating islet-derived 1 alpha (pancreatic stone protein, pancreatic threadprotein), PSPS1REG-1-alpha, PSPSMGC12447, PTPP19, regenerating islet-derived 1 alpha, Regenerating islet-derived protein 1-alpha, Regenerating protein I alpha, REGlithostathine 1 alpha
Reg1
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot
Unconjugated
RUO
P43137
5967
E. coli-derived recombinant mouse Reg1 Gln22-Gly165 Accession # P43137
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects mouse Reg1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) Reg2 is observed and less than 5% cross-reactivity with rmReg3A, recombinant human (rh) Reg1B, rhReg4, recombinant rat (rr) Reg3 and rrReg2 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only