missing translation for 'onlineSavingsMsg'
Learn More

LEC Refrigeration Refrigerator PGR273UK pharmacy, glass door 595mm x 634mm x 1570mm (w x d x h), 273L

Brand:  LEC Refrigeration PGR273UK

Refrigerator PGR273UK pharmacy, glass door 595mm x 634mm x 1570mm (w x d x h), 273L

Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 12948654

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.